Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD18 Mouse Monoclonal Antibody [Clone: MEM-48] (FITC (Fluorescein Isothiocyanate))
Anti-CD18 Mouse Monoclonal Antibody [Clone: MEM-48] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86014-100
Supplier:  Antibodies.com
New Product
Anti-CD18 Mouse Monoclonal Antibody [Clone: MEM-48] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA86014-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    95 subunit beta
  • Antigen symbol:
    CD18
  • Clonality:
    Monoclonal
  • Clone:
    MEM-48
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05107
  • Antigen synonyms:
    CD18|Complement component 3 receptor 3 and 4 subunit|Complement receptor C3 beta subunit|CD 18|Beta 2 (antigen CD18 (p95)|Cell surface adhesion glycoprotein LFA 1/CR3/P150,959 beta subunit precursor)|Cell surface adhesion glycoproteins LFA-1/CR3/p150|Complement receptor C3 subunit beta|Cell surface adhesion glycoproteins LFA 1/CR3/p150,95 subunit beta
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Leukocytes from a patient suffering from a LGL Type leukemia
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-48] antibody to CD18 (FITC) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD18
Clonality: Monoclonal
Clone: MEM-48
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human