Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Catalase Rabbit Polyclonal Antibody
Anti-Catalase Rabbit Polyclonal Antibody
Catalog # ANTIA92991-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Catalase Rabbit Polyclonal Antibody
Catalog # ANTIA92991-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cas1
  • Antigen symbol:
    Catalase
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04040
  • Antigen synonyms:
    Catalase|MGC138422|EC 1.11.1.6|Cs1|CAT|MGC138424|CATA_HUMAN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    60 kDa
  • Sequence:
    MADSRDPASDQMQHWKEQRAAQKADVLTTGAGNPVGDKLNVITVGPRGPLLVQDVVFTDEMAHFDRERIPERVVHAKGAGAFGYFEVTHDITKYSKAKVFEHIGKKTPIAVRFSTVAGESGSADTVRDPRGFAVKFYTEDGNWDLVGNNTPIFFIRDPILFPSFIHSQKRNPQTHLKDPDMVWDFWSLRPESLHQVSFLFSDRGIPDGHRHMNGYGSHTFKLVNA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1-225 of human Catalasealase (NP_001743.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Catalase for WB and IP with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IP
  • Recommended dilutions: WB: 1:1, 000 to 1:3, 000, IP: 1:20 to 1:50

Type: Primary
Antigen: Catalase
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat