Anti-beta Actin Rabbit Monoclonal Antibody [Clone: ARC5115-01]
Catalog # ANTIA92938-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:A26C1A
- Antigen symbol:beta Actin
- Clonality:Monoclonal
- Clone:ARC5115-01
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Zebrafish,Pig,Chicken,Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P60709
- Antigen synonyms:Actin, cytoplasmic 1, N-terminally processed|b actin|ACTB|A26C1B|Actx|Actin beta|ACTB_HUMAN|Actin cytoplasmic 1|beta Actin
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol, 0,05% BSA, and 0,05% Proclin 300
- Molecular weight:42 kDa
- Sequence:MDDDIAALVVDNGSGMCKAGFAGDDAPRAVFPSIVGRPRHQGVMVGMGQKDSYVGDEAQSKRGILTLKYPIEHGIVTNWDDMEKIWHHTFYNELRVAPEEHPVLLTEAPLNPKANREKMTQIMFETFNTPAMYVAIQAVLSLYASGRTTGIVMDSGDGVTHTVPIYEGYALPHAILRLDLAGRDLTDYLMKILTERGYSFTTTAEREIVRDIKEKLCYVALDFEQEMATAASSSSLEKSYELPDGQVITIGNERFRCPEALFQPSFLGMESCGIHETTFNSIMKCDVDIRKDLYANTVLSGGTTMYPGIADRMQKEITALAPSTMKIKIIAPPERKYSVWIGGSILASLSTFQQMWISKQEYDESGPSIVHRKCF
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human ß-actin
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit monoclonal [ARC5115-01] antibody to beta Actin for WB with samples derived from Human, Mouse, Rat, Chicken, Zebrafish and Porcine.
- Validated applications: WB
- Recommended dilutions: WB: 1:50, 000 to 1:200, 000
Type: Primary
Antigen: beta Actin
Clonality: Monoclonal
Clone: ARC5115-01
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Chicken, Zebrafish, Porcine