Anti-Cytokeratin 20 Mouse Recombinant Antibody [clone: rKRT20/6536]
ANTIA277911-100
New Product
- Antibody type:Primary
- Antigen name:CD20
- Antigen symbol:Cytokeratin 20
- Clonality:Recombinant
- Clone:rKRT20/6536
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P35900
- Antigen synonyms:Cytokeratin-20|Keratin|CK-20|CK20|CK 20|Cytokeratin20|KA20|K1C20_HUMAN|Cytokeratin 20
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 196-323, of human Cytokeratin 20. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rKRT20/6536] antibody to Cytokeratin 20 for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 20
Clonality: Recombinant
Clone: rKRT20/6536
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human