Anti-14-3-3 Theta + Tau Rabbit Polyclonal Antibody
ANTIA95555-100
New Product
- Antibody type:Primary
- Antigen name:14-3-3 protein T cell
- Antigen symbol:14-3-3 Theta + Tau
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P27348
- Antigen synonyms:14-3-3 protein T-cell|1433T_HUMAN|14-3-3 protein tau|Protein HS1|IC5|14-3-3 protein theta|14-3-3 Theta + Tau|YWHAQ tyrosine 3 monooxygenase/tryptophan 5-monooxygenase activation protein, theta polypeptide|Ywhaq
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:27 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human 14-3-3 thet (amino acids 41-90)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to 14-3-3 thet for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000
Type: Primary
Antigen: 14-3-3 Theta + Tau
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat