Order Entry
Czech Republic
ContactUsLinkComponent
Anti-BCAR1 Rabbit Polyclonal Antibody
Anti-BCAR1 Rabbit Polyclonal Antibody
Catalog # ANTIA95267-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-BCAR1 Rabbit Polyclonal Antibody
Catalog # ANTIA95267-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BCAR 1
  • Antigen symbol:
    BCAR1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P56945
  • Antigen synonyms:
    Breast cancer anti estrogen resistance 1|BCAR1_HUMAN|Cass1|Cas scaffolding protein family member 1|BCAR1|CAS|Breast cancer anti-estrogen resistance protein 1|Breast cancer anti estrogen resistance 1 protein|CAS1
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    93 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic phospho-peptide derived from human p130 Cas around the phosphorylation site of Tyr128
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to p130 Cas (phospho Tyr128) for WB and ELISA with samples derived from Human.

  • Validated applications: WB, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000

Type: Primary
Antigen: BCAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human