Anti-BCAR1 Rabbit Polyclonal Antibody
Catalog # ANTIA94599-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Druh protilátky:Primární
- Název antigenu:BCAR 1
- Symbol antigenu:BCAR1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Formulář:Liquid
- Identifikační číslo genu:UniprotID# P56945
- Synonyma antigenu:Breast cancer anti estrogen resistance 1|BCAR1_HUMAN|Cass1|Cas scaffolding protein family member 1|BCAR1|CAS|Breast cancer anti-estrogen resistance protein 1|Breast cancer anti estrogen resistance 1 protein|CAS1
- Skladovací pufr:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molekulární hmotnost:93 kDa
- Skladovací teplota:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Transportní teplota:Shipped on blue ice at +4 °C
- Imunogen:Synthetic phospho-peptide derived from human p130 Cas around the phosphorylation site of Tyr410
- Čištění:Purified from rabbit serum by antigen affinity chromatography using the immunising phospho peptide
- Typ balení:Plastic vial
- Bal.:100 µG
Specifications
About this item
Rabbit polyclonal antibody to p130 Cas (phospho Tyr410) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: BCAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat