Order Entry
Czech Republic
ContactUsLinkComponent
Anti-beta Actin Rabbit Monoclonal Antibody [clone: ARC5115-01]
Anti-beta Actin Rabbit Monoclonal Antibody [clone: ARC5115-01]
Katalogové číslo ANTIA309717-100
Supplier:  Antibodies.com
New Product
undefined
Anti-beta Actin Rabbit Monoclonal Antibody [clone: ARC5115-01]
Katalogové číslo ANTIA309717-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    A26C1A
  • Antigen symbol:
    beta Actin
  • Clonality:
    Monoclonal
  • Clone:
    ARC5115-01
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish,
    Pig,
    Chicken,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P60709
  • Antigen synonyms:
    Actin, cytoplasmic 1, N-terminally processed|b actin|ACTB|A26C1B|Actx|Actin beta|ACTB_HUMAN|Actin cytoplasmic 1|beta Actin
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,05% Proclin 300
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant protein of human ß-Actin.
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC5115-01] antibody to beta Actin for WB with samples derived from human, mouse, rat, chicken, zebrafish and porcine.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:10,000 to 1:100,000

Type: Primary
Antigen: beta Actin
Clonality: Monoclonal
Clone: ARC5115-01
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Chicken;Zebrafish;Porcine