Anti-Histone H2B (testis specific) Rabbit Polyclonal Antibody
Catalog # ANTIA309542-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Histone H2B (testis specific)
- Antigen symbol:Histone H2B (testis specific)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q96A08
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
- Molecular weight:20 kDa/15 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant protein of human Histone H2B (testis specific).
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Histone H2B (testis specific) for WB with samples derived from mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:100 to 1:500
Type: Primary
Antigen: Histone H2B (testis specific)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat