Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IP10 Rabbit Polyclonal Antibody
  ANTIA309104-100
New Product
undefined
Anti-IP10 Rabbit Polyclonal Antibody
  ANTIA309104-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    10 kDa interferon gamma induced protein
  • Antigen symbol:
    IP10
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02778
  • Antigen synonyms:
    CRG2|Chemokine CXC motif ligand 10|CXCL10(1-73)|C7|CXCL10|C X C motif chemokine 10|10 kDa interferon gamma-induced protein|Crg 2|Chemokine (C X C motif) ligand 10
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
  • Molecular weight:
    11 kDa
  • Sequence:
    VPLSRTVRCTCISISNQPVNPRSLEKLEIIPASQFCPRVEIIATMKKKGEKRCLNPESKAIKNLLKAVSKERSKRSP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 22-98 of human CXCL10/IP-10 (NP_001556.2).
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to IP10 for WB and ICC/IF with samples derived from human and mouse.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:1,000 to 1:5,000, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: IP10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse