Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EZH2 Mouse Monoclonal Antibody [clone: EZH2/4687]
Anti-EZH2 Mouse Monoclonal Antibody [clone: EZH2/4687]
Catalog # ANTIA314943-100
Supplier:  Antibodies.com
New Product
Anti-EZH2 Mouse Monoclonal Antibody [clone: EZH2/4687]
Catalog # ANTIA314943-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Enhancer of zeste 2
  • Antigen symbol:
    EZH2
  • Clonality:
    Monoclonal
  • Clone:
    EZH2/4687
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q15910
  • Antigen synonyms:
    Enhancer of zeste homolog 2 (Drosophila)|enhancer of zeste 2 polycomb repressive complex 2 subunit|Enx1h|ENX1|Enhancer of zeste, Drosophila, homolog 2|Enhancer of zeste homolog 2|ENX-1|Enx 1h|ENX 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    81 - 102 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment (around Amino Acids 300-550) of human EZH2 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [EZH2/4687] antibody to EZH2 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: EZH2
Clonality: Monoclonal
Clone: EZH2/4687
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human