Anti-Cytokeratin 19 Rabbit Recombinant Antibody [clone: KRT19/8091R]
Catalog # ANTIA314862-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:40 kDa keratin intermediate filament
- Antigen symbol:Cytokeratin 19
- Clonality:Recombinant
- Clone:KRT19/8091R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P08727
- Antigen synonyms:Cytokeratin-19|CK19|CK-19|K1CS|Keratin type I 40kD|Cytokeratin 19|CK 19|Keratin type I 40 kD|K1C19_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human KRT19 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRT19/8091R] antibody to Cytokeratin 19 for IHC-P with samples derived from human, mouse and rat.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 19
Clonality: Recombinant
Clone: KRT19/8091R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human, Mouse, Rat