Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8346R]
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8346R]
Catalog # ANTIA314859-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Cytokeratin 17 Rabbit Recombinant Antibody [clone: KRT17/8346R]
Catalog # ANTIA314859-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cytokeratin 17
  • Antigen symbol:
    Cytokeratin 17
  • Clonality:
    Recombinant
  • Clone:
    KRT17/8346R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q04695
  • Antigen synonyms:
    keratin 17 epitope S2|CK-17|Cytokeratin 17|Keratin|39.1|K1C17_HUMAN|keratin 17 epitope S1|Cytokeratin-17|K17|CK 17
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment of human KRT17 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRT17/8346R] antibody to Cytokeratin 17 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 17
Clonality: Recombinant
Clone: KRT17/8346R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human