Anti-Cytokeratin 15 Rabbit Recombinant Antibody [clone: KRT15/8312R]
ANTIA314857-100
New Product
- Antibody type:Primary
- Antigen name:AI528832
- Antigen symbol:Cytokeratin 15
- Clonality:Recombinant
- Clone:KRT15/8312R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P19012
- Antigen synonyms:Ka15|Cytokeratin 15|CK 15|K1C15_HUMAN|K1CO|Cytokeratin15|CK15|CK-15|Cytokeratin-15
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human full-length KRT15 protein
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [KRT15/8312R] antibody to Cytokeratin 15 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 15
Clonality: Recombinant
Clone: KRT15/8312R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human