Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cytokeratin 14 Rabbit Recombinant Antibody [clone: KRT14/7977R]
Anti-Cytokeratin 14 Rabbit Recombinant Antibody [clone: KRT14/7977R]
Catalog # ANTIA314854-100
Supplier:  ANTIBODIES.COM
New Product
Anti-Cytokeratin 14 Rabbit Recombinant Antibody [clone: KRT14/7977R]
Catalog # ANTIA314854-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CK 14
  • Antigen symbol:
    Cytokeratin 14
  • Clonality:
    Recombinant
  • Clone:
    KRT14/7977R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02533
  • Antigen synonyms:
    Cytokeratin14|Cytokeratin 14|Dowling Meara|Epidermolysis bullosa simplex|EBS4|CK-14|ck14|Cytokeratin-14|EBS3
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human KRT14 fragment (around Amino Acids 272-472). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRT14/7977R] antibody to Cytokeratin 14 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 14
Clonality: Recombinant
Clone: KRT14/7977R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human