Anti-Cytokeratin 14 Rabbit Recombinant Antibody [clone: KRT14/8691R]
Catalog # ANTIA314853-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CK 14
- Antigen symbol:Cytokeratin 14
- Clonality:Recombinant
- Clone:KRT14/8691R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P02533
- Antigen synonyms:Cytokeratin14|Cytokeratin 14|Dowling Meara|Epidermolysis bullosa simplex|EBS4|CK-14|ck14|Cytokeratin-14|EBS3
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant human KRT14 fragment (around Amino Acids 350-472). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [KRT14/8691R] antibody to Cytokeratin 14 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 14
Clonality: Recombinant
Clone: KRT14/8691R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human