Anti-Catalase Mouse Monoclonal Antibody [clone: HPYL/7225]
Catalog # ANTIA314564-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cas1
- Antigen symbol:Catalase
- Clonality:Monoclonal
- Clone:HPYL/7225
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:H. pylori
- Form:Liquid
- Antigen synonyms:Catalase|MGC138422|EC 1.11.1.6|Cs1|CAT|MGC138424|CATA_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:58 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant Helicobacter pylori Catalase protein fragment (around Amino Acids 323-445). The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HPYL/7225] antibody to Catalase for IHC-P with samples derived from Helicobacter pylori.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Catalase
Clonality: Monoclonal
Clone: HPYL/7225
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Helicobacter pylori