Anti-AMPK beta 1 Rabbit Polyclonal Antibody
ANTIA96781-100
New Product
- Antibody type:Primary
- Antigen name:1300015D22Rik
- Antigen symbol:AMPK beta 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9Y478
- Antigen synonyms:AMPK beta 1 chain|AMPK|5'-AMP-activated protein kinase beta-1 subunit|AMP-activated, noncatalytic, beta-1|5''-AMP-activated protein kinase subunit beta-1|AAKB1_HUMAN|AMP-activated protein kinase beta subunit|AMPK beta 1|AMP-ACTIVATED PROTEIN KINASE, NONCATALYTIC, BETA-1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:30 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human PRKAB1 (amino acids 10-59)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to PRKAB1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: AMPK beta 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat