Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Caspase 10 Rabbit Polyclonal Antibody
Anti-Caspase 10 Rabbit Polyclonal Antibody
Catalog # ANTIA96513-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Caspase 10 Rabbit Polyclonal Antibody
Catalog # ANTIA96513-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q92851-2,4
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    58 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human Caspase 10 (amino acids 430-479)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to Caspase 10 for WB, IHC, IF and ELISA with samples derived from Human.

  • Validated Applications: WB, IHC, IF, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human