Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43] (APC (Allophycocyanin))
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43] (APC (Allophycocyanin))
Catalog # ANTIA85983-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43] (APC (Allophycocyanin))
Catalog # ANTIA85983-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD59 antigen complement regulatory protein
  • Antigen symbol:
    CD59
  • Clonality:
    Monoclonal
  • Clone:
    MEM-43
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13987
  • Antigen synonyms:
    CD59 antigen|1F5|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|16.3A5|20 kDa homologous restriction factor|CD59 antigen p18 20|1F5 antigen
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Thymocytes and T lymphocytes
  • Purification:
    This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-43] antibody to CD59 (APC) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human