Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38]
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38]
Catalog # ANTIA85931-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38]
Catalog # ANTIA85931-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA DP
  • Antigen symbol:
    HLA DR + HLA DP
  • Clonality:
    Monoclonal
  • Clone:
    HL-38
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Pig,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    HLA DR + HLA DP|MHC Class II|HLA DR
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Raji Burkitt's lymphoma cell line
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [HL-38] antibody to HLA DR + HLA DP for IP, WB, IHC-P and flow cytometry with samples derived from human and porcine.

  • Validated applications: IP, WB, IHC-P, Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml

Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: HL-38
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine