Anti-HLA DR + HLA DP Mouse Monoclonal Antibody [Clone: HL-38] (FITC (Fluorescein Isothiocyanate))
ANTIA85925-100
New Product
- Antibody type:Primary
- Antigen name:HLA DP
- Antigen symbol:HLA DR + HLA DP
- Clonality:Monoclonal
- Clone:HL-38
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Pig,Human
- Form:Liquid
- Antigen synonyms:HLA DR + HLA DP|MHC Class II|HLA DR
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji Burkitt's lymphoma cell line
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [HL-38] antibody to HLA DR + HLA DP (FITC) for flow cytometry with samples derived from human and porcine.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: HLA DR + HLA DP
Clonality: Monoclonal
Clone: HL-38
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Porcine