Anti-CD45 Mouse Monoclonal Antibody [Clone: MEM-28] (Biotin)
ANTIA85893-100
New Product
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45
- Clonality:Monoclonal
- Clone:MEM-28
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes and T lymphocytes
- Tested applications:IHC-P, ICC
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [MEM-28] antibody to CD45 (Biotin) for flow cytometry, IP, WB, IHC-P and ICC with samples derived from human.
- Validated applications: Flow cytometry, IP, WB, IHC-P, ICC
- Recommended dilutions: Flow cytometry: 1 µg/ml
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human