Anti-CD45 Mouse Monoclonal Antibody [Clone: MEM-28] (PE (Phycoerythrin))
ANTIA85884-100
New Product
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45
- Clonality:Monoclonal
- Clone:MEM-28
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human thymocytes and T lymphocytes
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [MEM-28] antibody to CD45 (PE) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: MEM-28
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human