Anti-CD85j Mouse Monoclonal Antibody [Clone: GHI/75]
Catalog # ANTIA85831-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD85
- Antigen symbol:CD85j
- Clonality:Monoclonal
- Clone:GHI/75
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8NHL6
- Antigen synonyms:Ig like transcript 2|ILT-2|CD85j antigen|CD85 antigen|CD85 antigen like family member J|ILT2|CD85j|ILT 2|CD85 antigen-like family member J
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Hairy cell leukemia cells
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [GHI/75] antibody to CD85j for flow cytometry, IP and WB with samples derived from human.
- Validated applications: Flow cytometry, IP, WB
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD85j
Clonality: Monoclonal
Clone: GHI/75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human