Anti-CD85j Mouse Monoclonal Antibody [Clone: GHI/75] (PE (Phycoerythrin))
ANTIA85829-100
New Product
- Antibody type:Primary
- Antigen name:CD85
- Antigen symbol:CD85j
- Clonality:Monoclonal
- Clone:GHI/75
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q8NHL6
- Antigen synonyms:Ig like transcript 2|ILT-2|CD85j antigen|CD85 antigen|CD85 antigen like family member J|ILT2|CD85j|ILT 2|CD85 antigen-like family member J
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Hairy cell leukemia cells
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [GHI/75] antibody to CD85j (PE) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD85j
Clonality: Monoclonal
Clone: GHI/75
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human