Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-HLA DR Mouse Monoclonal Antibody [Clone: MEM-12]
  ANTIA85815-100
New Product
Anti-HLA DR Mouse Monoclonal Antibody [Clone: MEM-12]
  ANTIA85815-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    HLA DP
  • Antigen symbol:
    HLA DR
  • Clonality:
    Monoclonal
  • Clone:
    MEM-12
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    HLA DR + HLA DP|MHC Class II|HLA DR
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Thymocyte membrane
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [MEM-12] antibody to HLA DR for flow cytometry, IP and WB with samples derived from human.

  • Validated applications: Flow cytometry, IP, WB
  • Recommended dilutions: Flow cytometry: 2 µg/ml

Type: Primary
Antigen: HLA DR
Clonality: Monoclonal
Clone: MEM-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human