Anti-ICAM1 Mouse Monoclonal Antibody [Clone: 1H4] (Biotin)
ANTIA85720-100
New Product
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:ICAM1
- Clonality:Monoclonal
- Clone:1H4
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05362
- Antigen synonyms:Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji cells and spleen cells fused with NS1 cells
- Tested applications:IHC-P, IHC-Fr, ICC
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [1H4] antibody to ICAM1 (Biotin) for flow cytometry, WB, IHC-P, IHC-Fr and ICC with samples derived from human.
- Validated applications: Flow cytometry, WB, IHC-P, IHC-Fr, ICC
- Recommended dilutions: Flow cytometry: 2 to 5 µg/ml
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 1H4
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human