Order Entry
Czech Republic
ContactUsLinkComponent
Anti-alpha Fetoprotein Mouse Monoclonal Antibody [Clone: AFP-01]
Anti-alpha Fetoprotein Mouse Monoclonal Antibody [Clone: AFP-01]
Catalog # ANTIA85533-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-alpha Fetoprotein Mouse Monoclonal Antibody [Clone: AFP-01]
Catalog # ANTIA85533-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Afp
  • Antigen symbol:
    alpha Fetoprotein
  • Clonality:
    Monoclonal
  • Clone:
    AFP-01
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02771
  • Antigen synonyms:
    Alpha-fetoprotein|Alpha-fetoglobulin|Alpha fetoglobulin|AFPD|Alpha-1-fetoprotein|Alpha fetoprotein|Alpha fetoprotein precursor|alpha-fetoprotein, Hereditary persistence of, included|alpha 1 Fetoprotein
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Purified alpha Fetoprotein
  • Purification:
    Purified by precipitation and chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [AFP-01] antibody to alpha Fetoprotein for IP, ELISA and WB with samples derived from human.

  • Validated applications: IP, ELISA, WB

Type: Primary
Antigen: alpha Fetoprotein
Clonality: Monoclonal
Clone: AFP-01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human