Anti-HLA Class I Mouse Monoclonal Antibody [Clone: W6/32]
Catalog # ANTIA85491-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:DKFZp686P19218
- Antigen symbol:HLA Class I
- Clonality:Monoclonal
- Clone:W6/32
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Non-human primates,Human,Bovine,Cat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, without sodium azide (0,2 µm filter sterilised). Endotoxin level is less than 0,01 EU/µg of the protein, as determined by the LAL test
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Membrane of human tonsil cells
- Tested applications:IHC-Fr, ICC, CyTOF, Functional studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [W6/32] antibody to HLA Class I for flow cytometry, IP, WB, IHC-Fr, ICC, ELISA, CyTOF and functional studies with samples derived from human, bovine, feline and non-human primates.
- Validated applications: Flow cytometry, IP, WB, IHC-Fr, ICC, ELISA, CyTOF, functional studies
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Bovine, Cat, Non-Human Primates