Anti-HLA Class I Mouse Monoclonal Antibody [Clone: W6/32] (FITC (Fluorescein Isothiocyanate))
ANTIA85490-100
New Product
- Antibody type:Primary
- Antigen name:DKFZp686P19218
- Antigen symbol:HLA Class I
- Clonality:Monoclonal
- Clone:W6/32
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Non-human primates,Human,Bovine,Cat
- Form:Liquid
- Antigen synonyms:EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Membrane of human tonsil cells
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [W6/32] antibody to HLA Class I (FITC) for flow cytometry with samples derived from human, bovine, feline and non-human primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 3 µg/ml
Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: W6/32
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Bovine, Cat, Non-Human Primates