Anti-CD8 Mouse Monoclonal Antibody [Clone: MEM-31]
Catalog # ANTIA85485-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD8
- Clonality:Monoclonal
- Clone:MEM-31
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:OKT8 T cell antigen|CD8 antigen, alpha polypeptide (p32)|Leu2 T lymphocyte antigen|CD8A_HUMAN|CD8a|Leu2|MAL|CD8a antigen|CD8a molecule
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Crude thymus membrane fraction
- Tested applications:CyTOF
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-31] antibody to CD8 for flow cytometry, IP and CyTOF with samples derived from human.
- Validated applications: Flow cytometry, IP, CyTOF
- Recommended dilutions: Flow cytometry: 1 µg/ml
Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: MEM-31
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human