Order Entry
Czech Republic
ContactUsLinkComponent
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147] (PE (Phycoerythrin))
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147] (PE (Phycoerythrin))
Catalog # ANTIA85471-100
Supplier:  Antibodies.com
New Product
Anti-HLA Class I Mouse Monoclonal Antibody [Clone: MEM-147] (PE (Phycoerythrin))
Catalog # ANTIA85471-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp686P19218
  • Antigen symbol:
    HLA Class I
  • Clonality:
    Monoclonal
  • Clone:
    MEM-147
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    EA2.1|HLA Class I|HLA CDA12|HLA 5.4|HLA A|HLA 6.2|HLA B|EA1.2|HLA C
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Concentration:
    0,1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    PHA-activated peripheral blood lymphocytes
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-147] antibody to HLA Class I (PE) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/ml

Type: Primary
Antigen: HLA Class I
Clonality: Monoclonal
Clone: MEM-147
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human