Anti-PYROXD1 Goat Polyclonal Antibody
Catalog # ANTIA85088-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:DKFZp762G094
- Antigen symbol:PYROXD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PYROXD1|OTTHUMP00000240138|FLJ22028|Pyridine nucleotide-disulfide oxidoreductase domain-containing protein 1|OTTHUMP00000240140|PYROXD 1|PYRD1_HUMAN|Pyridine nucleotide disulphide oxidoreductase domain 1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:TCAEQLATHFPSED
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human PYROXD1 (N terminal)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to PYROXD1 for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: PYROXD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human