Anti-POLR3D Rabbit Polyclonal Antibody
Catalog # ANTIA99356-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AI326118
- Antigen symbol:POLR3D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05423
- Antigen synonyms:BN51|AW489084|DNA directed RNA polymerase III 47 kDa polypeptide|Polymerase (RNA) III (DNA directed) polypeptide D|POLR3D|BN51T|BN51 (BHK21) temperature sensitivity complementing|DNA directed RNA polymerase III subunit D|Polymerase (RNA) III (DNA directed) polypeptide D, 44kDa
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:44 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human RPC4 (amino acids 281-330)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to RPC4 for WB and ELISA with samples derived from human and mouse.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: POLR3D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse