Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Neurofilament Mouse Monoclonal Antibody [clone: ABT-NF]
Anti-Neurofilament Mouse Monoclonal Antibody [clone: ABT-NF]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-Neurofilament Mouse Monoclonal Antibody [clone: ABT-NF]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA98340-100
  • ANTIA98340-50
  • Antigen symbol:
  • Neurofilament
  • Neurofilament
  • Antigen name:
  • 150kDa medium
  • 150kDa medium
  • Antigen synonyms:
  • CMT2E|FLJ53642|CMT2CC|68 kDa neurofilament protein|160 kDa neurofilament protein|2 kDa neurofilament protein|AI847934|CMT1F|200 kDa neurofilament protein
  • CMT2E|FLJ53642|CMT2CC|68 kDa neurofilament protein|160 kDa neurofilament protein|2 kDa neurofilament protein|AI847934|CMT1F|200 kDa neurofilament protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • ABT-NF
  • ABT-NF
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P07196
  • UniprotID# P07196
  • Isotype:
  • IgG
  • IgG
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide derived from human Neurofilament
  • Synthetic peptide derived from human Neurofilament
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Immunogen affinity purified
  • Immunogen affinity purified
  • Storage buffer:
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC
  • IHC

Specifications

Frequently Bought Together