Order Entry
Czech Republic
ContactUsLinkComponent
Anti-AMPK gamma 1 Rabbit Polyclonal Antibody
Anti-AMPK gamma 1 Rabbit Polyclonal Antibody
Catalog # ANTIA306476-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-AMPK gamma 1 Rabbit Polyclonal Antibody
Catalog # ANTIA306476-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5' AMP activated protein kinase gamma 1 subunit
  • Antigen symbol:
    AMPK gamma 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P54619
  • Antigen synonyms:
    5''-AMP-activated protein kinase subunit gamma-1|AMPK subunit gamma-1|AMPKg|5' AMP activated protein kinase subunit gamma 1|AMPK gamma 1 chain|AAKG1_HUMAN|AMP activated protein kinase noncatalytic gamma 1 subunit|AMPK gamma 1|AMPK gamma1
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,05% Proclin 300
  • Molecular weight:
    37 kDa
  • Sequence:
    KSALVQIYELEEHKIETWREVYLQDSFKPLVCISPNASLFDAVSSLIRNKIHRLPVIDPESGNTLYILTHKRILKFLKLFITEFPKPEFMSKSLEELQIGT
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 100-200 of human AMPK?1 (NP_002724.1).
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to AMPK gamma 1 for WB with samples derived from Human and Rat.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:100 to 1:500

Type: Primary
Antigen: AMPK gamma 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat