Order Entry
Czech Republic
ContactUsLinkComponent
Anti-EAAT1 Rabbit Monoclonal Antibody [clone: ARC1714]
Anti-EAAT1 Rabbit Monoclonal Antibody [clone: ARC1714]
Catalog # ANTIA306133-100
Supplier:  Antibodies.com
New Product
undefined
Anti-EAAT1 Rabbit Monoclonal Antibody [clone: ARC1714]
Catalog # ANTIA306133-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    EA6
  • Antigen symbol:
    EAAT1
  • Clonality:
    Monoclonal
  • Clone:
    ARC1714
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P43003
  • Antigen synonyms:
    EAA1_HUMAN|GLAST|GLAST1|Excitatory amino acid transporter 1|GLAST-1|FLJ25094|glutamate/aspartate transporter, high affinity, sodium-dependent|EAAT1|Glial high affinity glutamate transporter
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    50 - 55 kDa/90 - 150 kDa/58 kDa/130 kDa
  • Sequence:
    GTKENMHREGKIVRVTAADAFLDLIRNMFPPNLVEACFKQFKTNYEKRSFKVPIQANETLVGAVINNVSEAMETLTRITEELVPVPGSVNGVNALGLVVFS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 150-250 of human EAAT1/SLC1A3 (P43003).
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC1714] antibody to EAAT1 for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type: Primary
Antigen: EAAT1
Clonality: Monoclonal
Clone: ARC1714
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat