Order Entry
Czech Republic
ContactUsLinkComponent
Anti-GOLPH2 Rabbit Monoclonal Antibody [clone: ARC0697]
Anti-GOLPH2 Rabbit Monoclonal Antibody [clone: ARC0697]
Catalog # ANTIA80822-100
Supplier:  Antibodies.com
New Product
undefined
Anti-GOLPH2 Rabbit Monoclonal Antibody [clone: ARC0697]
Catalog # ANTIA80822-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bA379P1.3
  • Antigen symbol:
    GOLPH2
  • Clonality:
    Monoclonal
  • Clone:
    ARC0697
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8NBJ4
  • Antigen synonyms:
    Golgi protein 73kD|Golgi protein 73 kD|C9orf155|GOLM1|Golgi membrane protein 1|Golgi membrane protein GP73|Chromosome 9 open reading frame 155|Golgi phosphoprotein 2|GOLM 1
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% Glycerol, 0,05% BSA, and 0,02% Sodium azide
  • Molecular weight:
    73 kDa
  • Sequence:
    VEKEETNEIQVVNEEPQRDRLPQEPGREQVVEDRPVGGRGFGGAGELGQTPQVQAALSVSQENPEMEGPERDQLVIPDGQEEEQEAAGEGRNQQKLRGEDD
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence within amino acids 250-350 of human GOLPH2 (Q8NBJ4)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit monoclonal [ARC0697] antibody to GOLPH2 for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:200

Type: Primary
Antigen: GOLPH2
Clonality: Monoclonal
Clone: ARC0697
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat