Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ATF6 Mouse Monoclonal Antibody [clone: 37-1]
Anti-ATF6 Mouse Monoclonal Antibody [clone: 37-1]
Catalog # ANTIA444-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ATF6 Mouse Monoclonal Antibody [clone: 37-1]
Catalog # ANTIA444-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Activating transcription factor 6
  • Antigen symbol:
    ATF6
  • Clonality:
    Monoclonal
  • Clone:
    37-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P18850
  • Antigen synonyms:
    cAMP-dependent transcription factor ATF-6 alpha|ATF6-alpha|ATF6A_HUMAN|alpha|ATF6 alpha|ATF6A|ATF6|ATF 6|cAMP dependent transcription factor ATF 6 alpha
  • Storage buffer:
    Supplied in phosphate buffered saline with 50% Glycerol
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant peptide corresponding to the N-terminal region of ATF6 alpha protein (GST tag)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal (37-1) antibody to ATF6 for WB and IP with samples derived from human and mouse.

  • Validated applications: WB, IP

Type: Primary
Antigen: ATF6
Clonality: Monoclonal
Clone: 37-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse