Anti-GAK Mouse Monoclonal Antibody [clone: 9-13]
ANTIA415-100
New Product
- Antibody type:Primary
- Antigen name:Auxilin 2
- Antigen symbol:GAK
- Clonality:Monoclonal
- Clone:9-13
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P97874
- Antigen synonyms:DNAJC26|Cyclin G associated kinase|EC 2.7.11.1|GAK protein|Cyclin-G-associated kinase|FLJ40395|FLJ16629|DNAJ26|GAK
- Storage buffer:Supplied in phosphate buffered saline with 50% Glycerol
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant rat GAK protein (a.a. 1-430)
- Pack type:Plastic vial
- Pk:100 µl
Mouse monoclonal (9-13) antibody to GAK for WB, IF and IP with samples derived from human and rat.
- Validated applications: WB, IF, IP
- Recommended dilutions: WB: 1:500, IF: 1:50
Type: Primary
Antigen: GAK
Clonality: Monoclonal
Clone: 9-13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat