Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD47 Recombinant Recombinant Antibody
Anti-CD47 Recombinant Recombinant Antibody
Catalog # ANTIA318955-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD47 Recombinant Recombinant Antibody
Catalog # ANTIA318955-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    CD47
  • Clonality:
    Recombinant
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Recombinant
  • Isotype:
    IgG4
  • Reactivity:
    Human
  • Form:
    Lyophilized
  • Gene ID:
    UniprotID# Q08722
  • Antigen synonyms:
    IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
  • Storage buffer:
    Lyophilised from sterile phosphate buffered saline, pH 7,4, Normally 5 - 8% trehalose is added as a protectant before lyophilisation
  • Storage temperature:
    Shipped at 4 °C. Lyophilized: Store at –20...–80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
  • Concentration:
    Reconstitution dependent
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant humanized monoclonal [Magrolimab Biosimilar] antibody to CD47 for ELISA and flow cytometry with samples derived from human.

  • Validated applications: ELISA, flow cytometry
  • Recommended dilutions: ELISA: 1:5000 to 10000, flow cytometry: 1:100

Type: Primary
Antigen: CD47
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG4
Reactivity: Human