ContactUsLinkComponent
PBS Tablet (Phosphate Buffered Saline Tablet)
PBS Tablet (Phosphate Buffered Saline Tablet)
Catalog # AATB10010
Supplier:  AAT Bioquest
New Product
CAS Number:  
undefined
PBS Tablet (Phosphate Buffered Saline Tablet)
Catalog # AATB10010
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    10 Tab.
  • Description:
    Phosphate buffered saline tablet
Specifications
About this item
Each PBS tablet prepares 100 mL of a 1X PBS solution containing 137 mM sodium chloride, 10 mM phosphate buffer, and 2.7 mM potassium chloride (pH 7.4).
  • Designed for ease of use
  • Saves time
PBS (phosphate buffered saline) is an isotonic buffer that closely mimics the human body's pH, osmolarity, and ion concentrations.
Caution: For research use only.