Zadání objednávky
Czech Republic
ContactUsLinkComponent
ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Catalog # AATB17258
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
undefined
ReadiUse™ dNTP Mix Set *10 mM PCR Grade*
Catalog # AATB17258
Supplier:  AAT BIOQUEST
New Product
CAS Number:  
Specifications
  • Bal.:
    1 mL
Specifications
About this item
10 mM dNTP Mix is a mixture of four nucleotides (dATP, dCTP, dGTP, dTTP) in purified water.
  • Each nucleotide is at a concentration of 10 mM
  • The formulation is stable for 12 months at −20 °C
10 mM dNTP mix is suitable for use in polymerase chain reaction (PCR), sequencing, fill-in reactions, nick translation, cDNA synthesis, and TdT-tailing reactions.
Caution: For research use only.