Anti-Bcl-2 Mouse Monoclonal Antibody [clone: 100/D5 + 124]
Catalog # ANTIA253049-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Apoptosis regulator Bcl 2
- Antigen symbol:Bcl-2
- Clonality:Monoclonal
- Clone:100/D5 + 124
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:Multiple
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P10415
- Antigen synonyms:C430015F12Rik|AW986256|Apoptosis regulator Bcl2|D830018M01Rik|BCL2_HUMAN|Apoptosis regulator Bcl-2|B cell CLL/lymphoma 2|D630044D05Rik|B cell leukemia/lymphoma 2
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:25 - 26 kDa
- Sequence:GAAPAPGIFSSQPG-Cys
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to amino acids 41 - 54 of human Bcl - 2 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [100/D5 + 124] antibody to Bcl-2 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Bcl-2
Clonality: Monoclonal
Clone: 100/D5 + 124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Multiple
Reactivity: Human