Anti-Cytokeratin 18 Mouse Recombinant Antibody [clone: rKRT18/1190]
Catalog # ANTIA252373-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Antigen symbol:Cytokeratin 18
- Clonality:Recombinant
- Clone:rKRT18/1190
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Mouse,Cat,Marmoset,Hamster,Pig,Horse,Sheep,Goat,Ferret,Human,Bovine,Dog,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:45 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant mouse monoclonal [rKRT18/1190] antibody to Cytokeratin 18 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Feline, Canine, Sheep, Goat, Horse and Marmoset Monkey.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Recombinant
Clone: rKRT18/1190
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Mouse;Rat;Hamster;Ferret;Bovine;Porcine;Cat;Dog;Sheep;Goat;Horse;Marmoset Monkey