Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
Catalog # ANTIA252311-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Cytokeratin 6A Mouse Recombinant Antibody [clone: rKRT6A/2100]
Catalog # ANTIA252311-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CK-6A
  • Antigen symbol:
    Cytokeratin 6A
  • Clonality:
    Recombinant
  • Clone:
    rKRT6A/2100
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02538
  • Antigen synonyms:
    K2C6A_HUMAN|Cytokeratin 6a|Keratin|CK6A|CK6C|Cytokeratin-6A|Cytokeratin-6D|CK-6D|CK6D
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    58 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human Cytokeratin 6A protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant mouse monoclonal [rKRT6A/2100] antibody to Cytokeratin 6A for IHC-P with samples derived from Human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 6A
Clonality: Recombinant
Clone: rKRT6A/2100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human