Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-APC Mouse Monoclonal Antibody [clone: ALi 12-28]
Catalog # ANTIA252049-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-APC Mouse Monoclonal Antibody [clone: ALi 12-28]
Catalog # ANTIA252049-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Adenomatous Polyposis Coli
  • Antigen symbol:
    APC
  • Clonality:
    Monoclonal
  • Clone:
    ALi 12-28
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P25054
  • Antigen synonyms:
    DP2.5|APC|FAP|APC_HUMAN|DP3|CC1|DP2|Deleted in polyposis 2.5|Adenomatous polyposis coli protein
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    66 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 1 - 433, of human APC protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [ALi 12-28] antibody to APC for ELISA, Flow Cytometry, IF and WB with samples derived from Human.

  • Validated applications: ELISA, Flow cytometry, IF, WB
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: APC
Clonality: Monoclonal
Clone: ALi 12-28
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human