Anti-HLA DRA Mouse Monoclonal Antibody [clone: 19-26.1]
Catalog # ANTIA252029-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:DR alpha chain
- Antigen symbol:HLA DRA
- Clonality:Monoclonal
- Clone:19-26.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01903
- Antigen synonyms:DR alpha chain precursor|HLA DR1B|DRB1|HLA DR3B|HLA class II histocompatibility antigen DR alpha chain|Histocompatibility antigen HLA DR alpha|DRA_HUMAN|HLA class II histocompatibility antigen|DRB4
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:36 kDa (alpha chain)/27 kDa (beta chain)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji cells
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [19-26.1] antibody to HLA DRA for Flow Cytometry, IF and IHC-P with samples derived from Human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: HLA DRA
Clonality: Monoclonal
Clone: 19-26.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human