Order Entry
Czech Republic
ContactUsLinkComponent
Anti-14-3-3 Sigma Mouse Monoclonal Antibody [clone: CPTC-SFN-2]
Anti-14-3-3 Sigma Mouse Monoclonal Antibody [clone: CPTC-SFN-2]
Catalog # ANTIA251928-100
Dodavatel:  Antibodies.com
New Product
undefined
Anti-14-3-3 Sigma Mouse Monoclonal Antibody [clone: CPTC-SFN-2]
Catalog # ANTIA251928-100
Dodavatel:  Antibodies.com
New Product

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    14 3 3 protein sigma
  • Antigen symbol:
    14-3-3 Sigma
  • Clonality:
    Monoclonal
  • Clone:
    CPTC-SFN-2
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P31947
  • Antigen synonyms:
    MGC143283|Mkrn3|1433S_HUMAN|14-3-3 sigma|Er|HME 1|Epithelial cell marker protein 1|14-3-3 protein sigma|HME1
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    28 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human SFN protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifikace

O tomto produktu

Mouse monoclonal [CPTC-SFN-2] antibody to 14-3-3 Sigma for WB and IHC-P with samples derived from Human.

  • Validated applications: WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: 14-3-3 Sigma
Clonality: Monoclonal
Clone: CPTC-SFN-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human